Learn More
Whirl-Pak™ Stand-Up Sample Bags
GREENER_CHOICE
Product Code. 11791796 Shop All Whirl-Pak Products
Change view
Click to view available options
Dimensions (L x W):
18.5 x 7.5 cm (7.25 x 3 in.)
23 x 11.5 cm (9 x 4.5 in.)
23 x 15 cm (9 x 6 in.)
38 x 15 cm (15 x 6 in.)
38 x 19 cm (15 x 7.5 in.)
Thickness (English):
2.50 mil
3 mil
4 mil
Capacity (Metric):
118 mL
532 mL
710 mL
1.24 L
2 L
Unit Size:
Pack of 250
Pack of 500
Pack of 6000
Product Code. Dimensions (L x W) Thickness (English) Capacity (Metric) unitSize
11791796 38 x 19 cm (15 x 7.5 in.) 4 mil 2 L Pack of 250
11716873 18.5 x 7.5 cm (7.25 x 3 in.) 2.50 mil 118 mL Pack of 500
30281139 18.5 x 7.5 cm (7.25 x 3 in.) 2.50 mil 118 mL Pack of 6000
11726873 23 x 11.5 cm (9 x 4.5 in.) 3 mil 532 mL Pack of 500
11766965 23 x 15 cm (9 x 6 in.) 3 mil 710 mL Pack of 500
11781796 38 x 15 cm (15 x 6 in.) 4 mil 1.24 L Pack of 250
6 options
Product Code. 11791796
Brand: Whirl-Pak™ B01451WA
Only null left
Add to basket
Add to basket
Stand-up bags make it very easy to add material to the bag and to remove sample for testing or analysis. The flat bottom ensures stability and the wire tabs hold the top open for easy access.
  • Paper-like finish virtually eliminates ink smearing
  • Surface that will accept a variety of writing instruments
  • Sterilized after manufacturing
Specifications
Description Whirl-Pak™ Stand-Up Bag
Autoclavable Not Autoclavable
Capacity (English) 69 oz.
Capacity (Metric) 2 L
Closure Type Tape and Wire
Color Clear with White Block
Disposable Single-use
Includes Bag
Material Polyethylene
Sterility Sterile
Thickness (Metric) 0.102 mm
Fill Volume 69 oz.
Latex Free Yes
No. per Pack 250/Pk.
Outer Packaging Cardboard
Protective Inner Packaging Plastic
Quantity 250/Pk.
Sterilization Method Ethylene Oxide Gas
Temperature Range (English) -320°F to +180°F
Temperature Range (Metric) -196°C to +82°C
Thickness (English) 4 mil
Dimensions (L x W) 38 x 19 cm (15 x 7.5 in.)
Length (English) 15 in.
Length (Metric) 38 cm
Width (English) 7.5 in.
Width (Metric) 19 cm
Product Type Stand-up Bag