Learn More
- Home
- Products
- Environmental Samplers
- General Purpose Sample Bags
- Whirl-Pak™ Standard Sample Bags
Whirl-Pak™ Standard Sample Bags
Product Code. p-32451
Shop All Whirl-Pak Products
Change view
Click to view available options
Dimensions (L x W):
12.5 x 6.5 cm
12.5 x 7.5 cm
18 x 9.5 cm
18.5 x 7.5 cm
18.5 x 7.5cm
19 x 13 cm
23 x 11.5 cm
23 x 11.5cm
23 x 15 cm
23 x 15cm
30 x 19cm
30.5 x 12.5 cm
38 x 12.5 cm
38 x 15 cm
38 x 19 cm
Thickness (English):
2.25 mil
2.5 mil
3 mil
4 mil
Capacity (English):
1 oz.
2 oz.
4 oz.
7 oz.
13 oz.
18 oz.
24 oz.
27 oz.
36 oz.
42 oz.
55 oz.
69 oz.
Product Type:
Sample Bag
Sample Bag with Labeling Area
Unit Size:
Pack of 10000
Pack of 500
20 product options available for selection
| Product Code. | Dimensions (L x W) | Thickness (English) | Capacity (English) | Product Type | unitSize |
|---|---|---|---|---|---|
| 11771664 | 12.5 x 7.5 cm | 2.25 mil | 2 oz. | Sample Bag | Pack of 500 |
| 11701973 | 18.5 x 7.5 cm | 2.25 mil | 4 oz. | Sample Bag | Pack of 500 |
| 11781664 | 18 x 9.5 cm | 3 mil | 7 oz. | Sample Bag | Pack of 500 |
| 11791664 | 19 x 13 cm | 3 mil | 13 oz. | Sample Bag | Pack of 500 |
| 11785714 | 23 x 11.5 cm | 2.5 mil | 18 oz. | Sample Bag | Pack of 500 |
| 11775774 | 23 x 15 cm | 3 mil | 24 oz. | Sample Bag | Pack of 500 |
| 11701674 | 23 x 15 cm | 4 mil | 24 oz. | Sample Bag | Pack of 500 |
| 11741684 | 30.5 x 12.5 cm | 3 mil | 27 oz. | Sample Bag | Pack of 500 |
| 11786508 | 38 x 12.5 cm | 3 mil | 36 oz. | Sample Bag | Pack of 500 |
| 11791963 | 38 x 15 cm | 3 mil | 42 oz. | Sample Bag | Pack of 500 |
| 11707093 | 38 x 19 cm | 3 mil | 69 oz. | Sample Bag | Pack of 500 |
| 11776965 | 12.5 x 6.5 cm | 2.25 mil | 1 oz. | Sample Bag | Pack of 500 |
| 11781963 | 12.5 x 7.5 cm | 2.25 mil | 2 oz. | Sample Bag with Labeling Area | Pack of 500 |
| 11735804 | 18.5 x 7.5cm | 2.25 mil | 4 oz. | Sample Bag with Labeling Area | Pack of 500 |
| 11771963 | 23 x 11.5cm | 2.5 mil | 18 oz. | Sample Bag with Labeling Area | Pack of 500 |
| 11761684 | 23 x 15 cm | 4 mil | 24 oz. | Sample Bag with Labeling Area | Pack of 500 |
| 11771684 | 23 x 15cm | 3 mil | 24 oz. | Sample Bag with Labeling Area | Pack of 500 |
| 11734834 | 30.5 x 12.5 cm | 3 mil | 27 oz. | Sample Bag with Labeling Area | Pack of 500 |
| 11751684 | 30 x 19cm | 4 mil | 55 oz. | Sample Bag with Labeling Area | Pack of 500 |
| 30286575 | 18.5 x 7.5 cm | 2.25 mil | 4 oz. | Sample Bag with Labeling Area | Pack of 10000 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Code produit.
11771664
missing translation for 'mfr'
Whirl-Pak™
missing translation for 'supplierNo'
B01009
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Features puncture proof tabs for protection from damage due to wire-end protrusion. Easy-to-close bags are ideal for sampling and product analysis. Available in plain or write-on styles.
- Transparent polyethylene bags
- Whirling or folding thebag over three times and securing with the closure tabs creates a leak-proof sampling container
- Gas sterilized, with documentation available
- Pull tabs help prevent contamination from finger contact when opening
- Write-on bags have area that accepts ballpoint pen or pencil
- Bags can be frozen to any temperature; including liquid nitrogen--careful handling required after freezing
- Thickness, 2.25 to 4 mil
- RNase, DNase, pryogen and BPH free
- Comply with regulatory and industrial requirements of EPA, FDA, HACCP and USDA
- Bags should not be used at temperatures above 180°F (82°C)
Spécification
Spécification
| Description | Plain Style |
| Autoclavable | Not Autoclavable |
| Closure Type | Closing Tab |
| Color | Clear |
| Disposable | Single-use |
| Material | Polyethylene |
| Sterility | Sterile |
| Latex Free | Yes |
| No. per Pack | 500/Pk. |
| Outer Packaging | Cardboard |
| Protective Inner Packaging | Plastic |
| Quantity | 500/Pk. |
| Sterilization Method | Ethylene Oxide Gas |
| Temperature Range (English) | -320°F to +180°F |
| Temperature Range (Metric) | -196°C to +82°C |
| Thickness (English) | 2.25 mil |
| Dimensions (L x W) | 12.5 x 7.5 cm |
| Length (English) | 5 in. |
| Width (English) | 3 in. |
| Product Type | Sample Bag |
| Capacity (English) | 2 oz. |
| Capacity (Metric) | 58 mL |
| Length (Metric) | 12.5 cm |
| Width (Metric) | 7.5 cm |
| Thickness (Metric) | 0.057 mm |
| Afficher plus Afficher moins |
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?