Learn More
Sampling Systems™ Narrow Sampler Cleaning Brushes
Product Code. 15849411 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
600 mm
1100 mm
1600 mm
2100 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
15849411 2100 mm Each
15142818 1100 mm Each
15839411 1600 mm Each
15859411 600 mm Each
4 options
Product Code. 15849411
Brand: Sampling Systems™ 8020N2100
Only null left
Add to basket
Add to basket
Ensure that the sampler is properly cleaned in order to prevent cross contamination. Sampling Systems™ Narrow Sampler Cleaning Brushes are a range of brushes that are ideal for cleaning both stainless steel and plastic samplers.
  • Hygienic nylon brushes
  • Robust aluminum handle
  • Narrow brush shape
Specifications
Product Type Cleaning Brush
Handle Material Aluminum
Tip Type Narrow
Diameter (Metric) Bristle 14 mm
For Use With (Equipment) Tubes with 7 to 14mm Diameter
Length (Metric) 2100 mm