Description
- Hygienic nylon brushes
- Robust aluminum handle
- Narrow brush shape
Specifications
Specifications
| Product Type | Cleaning Brush |
| Handle Material | Aluminum |
| Tip Type | Narrow |
| Diameter (Metric) Bristle | 14 mm |
| For Use With (Equipment) | Tubes with 7 to 14mm Diameter |
| Length (Metric) | 1100 mm |