Learn More
Kautex™ Narrow Neck LDPE Bottles, Natural
Product Code. 15951381
Change view
Click to view available options
Capacity (Metric):
3 mL
10 mL
20 mL
50 mL
100 mL
200 mL
250 mL
1000 mL
Unit Size:
Each
Pack of 1500
Pack of 1800
Pack of 3600
Product Code. Capacity (Metric) unitSize
15951381 20 mL Pack of 1800
17898117 3 mL Pack of 1500
15941381 10 mL Pack of 3600
10292271 50 mL Each
10473331 100 mL Each
10057990 200 mL Each
10393871 250 mL Each
10444461 1000 mL Each
8 options
Product Code. 15951381
Brand: Kautex™ 2000070501
Use Kautex™ Narrow Neck LDPE Bottles, Natural in the laboratory as an essential tool with which to conduct research and experiments. Secure them tightly with closures available for purchase separately.
  • Without closure
  • Narrow mouth
  • LDPE construction
  • Metric capacity
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 20 mL
Material LDPE
No. per Pack 1 Ea.
Packaging Bottle
Quantity 1 Ea.
Mouth Narrow
Height (Metric) 56.5 mm
Product Type Bottle
Diameter (Metric) 31.5 mm