Learn More
Kautex™ Narrow Neck LDPE Bottles, Natural
Product Code. 10444461
Click to view available options
Capacity (Metric):
3 mL
10 mL
20 mL
50 mL
100 mL
200 mL
250 mL
1000 mL
Unit Size:
Each
Pack of 1500
Pack of 1800
Pack of 3600
Product Code. 10444461
Brand: Kautex™ 2000070508
Only null left
Add to basket
Add to basket
Use Kautex™ Narrow Neck LDPE Bottles, Natural in the laboratory as an essential tool with which to conduct research and experiments. Secure them tightly with closures available for purchase separately.
  • Without closure
  • Narrow mouth
  • LDPE construction
  • Metric capacity
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Material LDPE
No. per Pack 1 Ea.
Packaging Bottle
Quantity 1 Ea.
Mouth Narrow
Height (Metric) 223.5 mm
Product Type Narrow Mouth Bottle
Diameter (Metric) 97.5 mm