Learn More
Gravis Narrow Neck Clear Glass Bottles
Description
- Available in a range of capacities
- Manufactured from soda lime glass
- Caps not included
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Color | Clear |
| Diameter (Metric) Inner | 24 mm |
| Material | Soda Lime Glass |
| Neck ID (Metric) | 18 |
| No. per Pack | 195Pack |
| Mouth | Narrow |
| Height (Metric) | 51 mm |
| Product Type | Storage Bottle |