Learn More
Naratriptan 2-Carboxylic Acid, TRC
Product Code. 30477511
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477511 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30477511 Supplier Toronto Research Chemicals Supplier No. TRCN3787452.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Naratriptan 2-Carboxylic Acid
CAS 874583-40-1
Molecular Formula C18H25N3O4S
Quantity 2.5 mg
Synonym 3-(1-Methyl-4-piperidyl)-5-[2-(methylsulfamoyl)ethyl]-1H-indole-2-carboxylic Acid
InChI Formula InChI=1S/C18H25N3O4S/c1-19-26(24,25)10-7-12-3-4-15-14(11-12)16(17(20-15)18(22)23)13-5-8-21(2)9-6-13/h3-4,11,13,19-20H,5-10H2,1-2H3,(H,22,23)
SMILES CNS(=O)(=O)CCc1ccc2[nH]c(C(=O)O)c(C3CCN(C)CC3)c2c1
IUPAC Name 3-(1-methylpiperidin-4-yl)-5-[2-(methylsulfamoyl)ethyl]-1H-indole-2-carboxylic acid
Molecular Weight (g/mol) 379.47
Formula Weight 379.1566