Learn More
NAPSA Monoclonal Antibody (OTI3G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813601
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
NAPSA Monoclonal antibody specifically detects NAPSA in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| NAPSA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O96009 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 9476 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| OTI3G2 | |
| Unconjugated | |
| NAPSA | |
| asp 4; ASP4; Aspartyl protease 4; KAP; Kdap; KDAP-1; kidney-derived aspartic protease-like protein; Nap; NAP1; NAPA; Napsa; napsin A aspartic peptidase; napsin-1; Napsin-A; pronapsin; pronapsin A; SNAPA; TA01/TA02 | |
| Human recombinant protein fragment corresponding to amino acids 64-244 of human NAPSA (NP_004842) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.