missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NAPRT1 (Human) Recombinant Protein

Human NAPRT1 full-length ORF ( NP_660202.2, 1 a.a. - 466 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00093100-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136803

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MALGYWRAGRARDAAEFELFFRRCPFGGAFALAAGLRDCVRFLRAFRLRDADVQFLASVLPPDTDPAFFEHLRALDCSEVTVRALPEGSLAFPGVPLLQVSGPLLVVQLLETPLLCLVSYASLVATNAARLRLIAGPEKRLLEMGLRRAQGPDGGLTASTYSYLGGFDSSSNVLAGQLRGVPVAGTLAHSFVTSFSGSEVPPDPMLAPAAGEGPGVDLAAKAQVWLEQVCAHLGLGVQEPHPGERAAFVAYALAFPRAFQGLLDTYSVWRSGLPNFLAVALALGELGYRAVGVRLDSGDLLQQAQEIRKVFRAAAAQFQVPWLESVLIVVSNNIDEEALARLAQEGSEVNVIGIGTSVVTCPQQPSLGGVYKLVAVGGQPRMKLTEDPEKQTLPGSKAAFRLLGSDGSPLMDMLQLAEEPVPQAGQELRVWPPGAQEPCTVRPAQVEPLLRLCLQQGQVAAPPPSS
Specifications

Specifications

NP_660202.2
nicotinate phosphoribosyltransferase domain containing 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NAPRT1
GST
93100
Wheat germ expression system
25 μg
PP3856
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers