missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15604-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
NAPG is soluble NSF attachment protein gamma. The soluble NSF attachment proteins (SNAPs) enable N-ethyl-maleimide-sensitive fusion protein (NSF) to bind to target membranes. NSF and SNAPs appear to be general components of the intracellular membrane fusion apparatus, and their action at specific sites of fusion must be controlled by SNAP receptors particular to the membranes being fused. The product of this gene mediates platelet exocytosis and controls the membrane fusion events of this process.Specifications
NAPG | |
Polyclonal | |
Unconjugated | |
NAPG | |
GAMMASNAP, NAPG, SNAP gamma, SNAPG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108123, 307382, 8774 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99747, Q9CWZ7 | |
NAPG | |
NAPG Fusion Protein Ag7988 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |