missing translation for 'onlineSavingsMsg'
Learn More

Napabucasin, MedChemExpress

Product Code. 16421116
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16421116

Brand: Medchem Express HY13919

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Napabucasin (BBI608) is a STAT3 inhibitor which blocks stem cell activity in cancer cells.

Napabucasin (BBI608) is a STAT3 inhibitor which blocks stem cell activity in cancer cells.

Specifications

Chemical Name or Material Napabucasin
Physical Form Powder
CAS 83280-65-3
Color Green Yellow
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C14H8O4
For Use With (Application) Cancer-Kinase/protease
Synonym BBI608
Solubility Information DMSO : 4.44 mg/mL (18.48 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C2OC(C(C)=O)=C1)C3=C(C2=O)C=CC=C3
Molecular Weight (g/mol) 240.21
Formula Weight 240.21
Percent Purity 99.27%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.