missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAP1L4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16018-1-AP-150UL
This item is not returnable.
View return policy
Description
NAP1L4 acts as histone chaperone in nucleosome assembly.Specifications
| NAP1L4 | |
| Polyclonal | |
| Unconjugated | |
| NAP1L4 | |
| hNAP2, NAP 2, NAP1L4, NAP2, NAP2L, Nucleosome assembly protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17955, 361684, 4676 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5U2Z3, Q78ZA7, Q99733 | |
| Nap1l4 | |
| NAP1L4 Fusion Protein Ag8827 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction