Learn More
Trajan Scientific and Medical™ NanoVolume Syringes
Product Code. 15298433
Click to view available options
Volume (Metric):
0.5 μL
Unit Size:
Each
Product Code. 15298433
Brand: Trajan Scientific and Medical™ 000353
Designed to inject down to 50nL with high precision and accuracy.
  • High precision and accuracy for small volume injections
  • Replacement plunger/needle kits
  • Kits are matched and must be replaced as a set
Specifications
Description NanoVolume Syringes
Needle Gauge 26 G
Diameter (Metric) Inner Needle 0.155 mm
Diameter (Metric) Outer Needle 0.47 mm
Length (Metric) 50 mm
Volume (Metric) 0.5 μL
Needle Point Style Cone