Learn More
Implen™ NanoPhotometer™ IQ/OQ Documentation and Standard Solution
Product Code. p-7214885 Shop All Implen Products
Click to view available options
For Use With (Equipment):
NanoPhotometer N120
NanoPhotometer N60
Unit Size:
Each
Product Code. 15533214
Brand: Implen™ N60Q
In compliance with international standards in regulated environments, documented verification is provided with Installation Qualification and Operation Qualification (IQ/OQ) Package Conforming Data to verify that instruments are working according to specifications in case of an audit.
  • Documentation provided on a USB flash drive as an Excel file
  • Includes NanoPhotometer™ appropriate standard solution (N-568-S or N-120-S)
Specifications
For Use With (Equipment) NanoPhotometer N60
Certifications/Compliance According to Internal SOP
Calibration Type Self-calibration
Product Type IQ/OQ Package Conforming Data
Product Line NanoPhotometer
Includes IQ/OQ Documentation and Standard Solution (N-568-S)
For Research Use Only