Learn More
NANOG Mouse anti-Human, Unconjugated, Clone: 3A2E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67255-1-IG-20UL
This item is not returnable.
View return policy
Description
The NANOG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NANOG. This antibody recognizes human, mouse antigen. The NANOG antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NANOG | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot | |
| 3A2E1 | |
| Unconjugated | |
| 20 μL | |
| 79923 | |
| IgG2a |