Learn More
NANOG Mouse anti-Human, Unconjugated, Clone: 3A2E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67255-1-IG-20UL
 View more versions of this product
Product Code. 16809785
  • 1710.33 NOK / 20µL
Description
The NANOG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NANOG. This antibody recognizes human, mouse antigen. The NANOG antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NANOG
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse
Western Blot
3A2E1
Unconjugated
20 μL
79923
IgG2a