missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NANOCOLOR™ COD Tube Tests

Easy, fast COD measurements that conform to ISO 15705 standards. Macherey-Nagel™ Nanocolor™ COD Tube Tests include exactly pre-dosed reagents in 16mm cuvettes and additional reagents for easy handling, high safety and maximum precision. Available in a range of measurement ranges.

Brand:  Macherey-Nagel™ 985012

Additional Details : Weight : 0.20000kg Transport : UN number : 3316 Chem class : 9 Pack group : II


 View more versions of this product

Product Code. 11390625

  • 980.00 NOK / 1 test
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

Nanocolor™ tube tests feature easy handling for routine, laboratory and process analysis. They are ideally packed in stable boxes with color coded labels, giving all relevant information at a glance. A range of standardized tube tests are available for ISO 15705-conforming COD analysis using tube tests that are suitable for photometric evaluation. Each tube test is bar coded for automatic recognition by Macherey-Nagel photometers.

  • Easy
    • Colored pictograms as step-by-step instruction
    • Big cuvettes for easy pipetting
    • Barcoded cuvettes for automatic test selection
  • Safe
    • Convenient withdrawal of tubes from the box
    • No contact with chemicals
    • Reactions based on internationally accepted standard methods
  • Reliable
    • Precisely pre-dosed reagents
    • Adequate test for every application
    • Constant high quality from batch to batch
Specifications

Specifications

ISO 6060
Determination of chemical oxygen demand (COD)
20 x 16mm Cuvettes, pre-filled with reagent and one null cuvette
1 Each
Potassium dichromate
Cuvettes
Macherey-Nagel Photometers
20
5.0 to 60g/L O2
NANOCOLOR Tube Test COD
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • NANOCOLOR COD 60000

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H413-May cause long lasting harmful effects to aquatic life.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : silver sulphate, sulphuric acid, mercury(II) sulphate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers