Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 750, Chromotek
Product Code. 18015125
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
Product Code. Quantity unitSize
18015125 100 μL 100µL
18099815 10 μL 10µL
2 options
Product Code. 18015125
Brand: Chromotek shuGCL7502100
Alpaca Recombinant Monoclonal Antibody
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Specifications
Antigen Nano-Secondary™
Conjugate CoraLite Plus 750
Host Species Alpaca
Quantity 100 μL
Primary or Secondary Secondary
Target Species Human
Form Liquid
Isotype IgG