Learn More
NAMPT/PBEF Mouse anti-Human, Mouse, Rat, Clone: 3D4D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66385-1-IG-20UL
This item is not returnable.
View return policy
Description
Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.Specifications
| NAMPT/PBEF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P43490, Q80Z29, Q99KQ4 | |
| Nampt | |
| NAMPT/PBEF Fusion Protein Ag2434 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| 3D4D8 | |
| Unconjugated | |
| NAMPT | |
| 1110035O14Rik, NAmPRTase, NAMPT, PBEF, PBEF1, Pre B cell enhancing factor, VF, VISFATIN | |
| Mouse | |
| Protein A | |
| RUO | |
| 10135, 297508, 59027 | |
| -20°C | |
| Liquid |