missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NALP6 Antibody (920631) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9145JF635
This item is not returnable.
View return policy
Beskrivelse
NALP6 Monoclonal antibody specifically detects NALP6 in Human samples. It is validated for Flow Cytometry,CyTOF
Tekniske data
| NALP6 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5 | |
| E.coli-derived recombinant human NLRP6/NALP6, Lys35-Arg193, Accession # P59044 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 920631 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology, Inflammation | |
| 171389 | |
| Store at 4°C in the dark. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion