missing translation for 'onlineSavingsMsg'
Learn More

NALP12 Antibody, FITC, Novus Biologicals™

Product Code. 18510121 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
18510121 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 18510121

Brand: Novus Biologicals NBP176293F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NALP12 Polyclonal antibody specifically detects NALP12 in Human, Mouse, Rat, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP12
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias FCAS2, Monarch-1, NACHT, leucine rich repeat and PYD containing 12, NACHT, LRR and PYD domains-containing protein 12, NALP12LRR and PYD containing protein 12, NLR family, pyrin domain containing 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 12, PYPAF7Monarch1, PYRIN-containing APAF1-like protein 7, Regulated by nitric oxide, RNO2
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human NALP12. The immunogen is located within amino acids 110 - 160 of NALP12. Amino Acid Squence: TPRKDPQETYRDYVRRKF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 91662
Target Species Human, Mouse, Rat, Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.