Learn More
Nalidixic acid sodium salt, 98.1%, MP Biomedicals™
Product Code. 11412462 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412462 1 g 1g
11422462 5 g 5g
11432462 25 g 25g
11442462 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11412462 Supplier MP Biomedicals Supplier No. 0215172401
  • ONLINE EXCLUSIVE PRICE
    378.00 NOK
    359.17 NOK / 1g
    Save 18.83 NOK
    4% Off
 Restricted Item
Estimated Shipment: 09-09-2026
Chemical Identifiers
CAS 389-08-2
Molecular Formula C12H12N2O3
Molecular Weight (g/mol) 232.239
InChI Key MHWLWQUZZRMNGJ-UHFFFAOYSA-N
Synonym nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
PubChem CID 4421
ChEBI CHEBI:100147
IUPAC Name 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
Specifications
Color White
Assay Percent Range ∽ 98%
Quantity 1 g
Formula Weight 254.2
Physical Form Powder
Chemical Name or Material Nalidixic Acid Sodium Salt