Learn More
Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
GREENER_CHOICE
Product Code. p-4520875
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
Array:
3 x 8
4 x 10
6 x 12
For Use With (Equipment):
13mm Tubes
16mm Tubes
20mm Tubes
25mm Tubes
30mm Tubes
Unit Size:
Each
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Array For Use With (Equipment) unitSize
10603161 Red 6 x 12 13mm Tubes Each
10149970 Blue 6 x 12 13mm Tubes Each
10180351 Green 6 x 12 13mm Tubes Each
10333461 Yellow 6 x 12 13mm Tubes Each
10634481 Orange 6 x 12 13mm Tubes Each
10693151 White 6 x 12 13mm Tubes Each
10435071 Red 6 x 12 16mm Tubes Each
10098210 Blue 6 x 12 16mm Tubes Each
10495061 Green 6 x 12 16mm Tubes Each
10343461 Yellow 6 x 12 16mm Tubes Each
10717961 Orange 6 x 12 16mm Tubes Each
10281761 White 6 x 12 16mm Tubes Each
10519081 Red 4 x 10 20mm Tubes Each
10323701 Blue 4 x 10 20mm Tubes Each
10769491 Green 4 x 10 20mm Tubes Each
10604101 Yellow 4 x 10 20mm Tubes Each
10169970 Orange 4 x 10 20mm Tubes Each
10291761 White 4 x 10 20mm Tubes Each
10663721 Red 4 x 10 25mm Tubes Each
10415411 Blue 4 x 10 25mm Tubes Each
10231671 Green 4 x 10 25mm Tubes Each
10602401 Yellow 4 x 10 25mm Tubes Each
10738151 Orange 4 x 10 25mm Tubes Each
10474101 White 4 x 10 25mm Tubes Each
10241861 Red 3 x 8 30mm Tubes Each
10778721 Blue 3 x 8 30mm Tubes Each
10602971 Green 3 x 8 30mm Tubes Each
10038180 Yellow 3 x 8 30mm Tubes Each
10242101 Orange 3 x 8 30mm Tubes Each
10644101 White 3 x 8 30mm Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 10603161 Supplier Thermo Scientific™ Supplier No. 59700513
Add to basket
Add to basket
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with these strong, durable racks.
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks. Molded in a single piece for exceptional strength and durability, these racks have no coating to chip, peel or rust.
  • Smooth finish leaves fewer places for contaminants to lodge; no welds to break, no sharp edges.
  • Endplates accept large labels for easy identification of samples.
  • Higher specific gravity than water so it stays submerged in a water bath and won't float.
  • Withstands most lab chemicals and temperatures up to 121°C for a variety of lab procedures.
  • Available in six bright colors to help keep samples organized.
  • 15mL conical tubes fit 20mm racks; 50mL conical fit 30mm racks.
  • No assembly is required; rack is ready to use.
TRUSTED_SUSTAINABILITY
Specifications
Description Test Tube Rack Unwire™ Red 13mm
Autoclavable Autoclavable
Color Red
Holds Tubes (13 mm)
Material RESMER
Positions 72
Shape Rectangular
For Use With (Equipment) 13mm Tubes
Dimensions (L x W x H) 7.75 x 4 x 2.25 in. (20 x 10.2 x 5.7 cm)
Length (Metric) 20 cm
Length (English) 7.75 in.
Width (English) 4 in.
Height (English) 2.25 in.
Diameter (Metric) Opening 13 mm
Product Type Test Tube Rack
Diameter (Metric) Inner 13 mm
Shelves 12
Height (Metric) 5.7 cm
Array 6 x 12
Width (Metric) 10.2 cm
Show More Show Less
Which Unwire racks are autoclavable?
Any of our ResMer polyketone test tube racks can be autoclaved, by our standard validated parameters (121 degrees Celsius at 15 psig for 20 minutes). Due to their molded structure, our polypropylene Unwire racks (unlike our other polyproylene labware products) cannot be autoclaved.
Which of your Unwire test tube racks can be used for 15 mL or 50 mL sterile conical centrifuge tubes?
Any of our Unwire racks with spaces for tubes 20 mm and 30 mm in diameter can be used, respectively, for 15 mL and 50 mL sterile conical centrifuge tubes.