Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures
Product Code. 10232673
Change view
Click to view available options
Closure Size:
20 to 415 for serum vial
24 to 415 modified
28 to 415 modified
38 to 415 modified
38 to 430 modified
43 to 415 modified
53 to 415 modified
53B
83B
Unit Size:
Each
Product Code. Closure Size unitSize
10232673 83B Each
11369754 20 to 415 for serum vial Each
10189792 20 to 415 for serum vial Each
10180703 24 to 415 modified Each
10190703 28 to 415 modified Each
11359754 38 to 415 modified Each
10571845 43 to 415 modified Each
10405323 53 to 415 modified Each
10100903 53B Each
10242673 38 to 430 modified Each
10 options
Product Code. 10232673
Brand: Thermo Scientific™ 21951183
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Choose these torque wrench fittings designed for specific HDPE closures to achieve accurate torque readings when applying closures onto bottles.
Choose Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures to achieve accurate torque readings when applying closures onto bottles. Proper torquing of Nalgene closures is critical to leakproof† sealing.
  • Epoxy
  • Specifically designed for Nalgene HDPE closures
Compatible with:
Thermo Scientific Nalgene Torque Wrenches (2195-6169 and -6177) † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fittings
Closure Size 83B
Drive Size 3/8 in.
For Use With (Equipment) Nalgene Torque Wrenches (Cat. Nos. 2195-6169 and -6177)
Material Epoxy