Learn More
Thermo Scientific™ Nalgene™ Stopcocks
Product Code. 15991651
Change view
Click to view available options
Plug Bore:
2 mm
4 mm
Unit Size:
Each
Pack of 6
Product Code. Plug Bore unitSize
15991651 2 mm Pack of 6
11369554 2 mm Pack of 6
15223079 4 mm Pack of 6
10279070 4 mm Each
4 options
Product Code. 15991651
Brand: Thermo Scientific™ 64600002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Develop in-house fluid transfer systems for accurate filling or dispensing. These stopcocks provide reliable, accurate liquid flow control.
Permit reliable, accurate liquid flow control in liquid handling systems by using Thermo Scientific™ Nalgene™ Stopcocks. They are ideal for use in developing in-house fluid transfer systems for one-way fluid path for accurate filling or dispensing. The two-way stopcocks feature serrated tubulation on each end.
  • Serrated tubulation on each end accepts 1/2″ through 5/16″ tubing
  • Polypropylene with Teflon™ TFE plug
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Type Stopcock
Plug Bore 2 mm
Material Polypropylene
No. per Pack 1
No. per Case 6
For Use With (Equipment) In-House Fluid Transfer System
Plug Material Polypropylene
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.