Learn More
Thermo Scientific™ Nalgene™ Square Polycarbonate Graduated Bottles without Closure: Tray Pack
Description
Choose Thermo Scientific™ Nalgene Square Polycarbonate Graduated Bottles for storage of aqueous solutions and short-term storage of media. These bottles are durable, autoclavable and feature an easy-to-handle square design.
- Transparent
- Easy-to-handle square design
- Autoclavable
- Tough, durable even at low temperatures
- With molded-in graduations and drip-proof neck
Ordering Information:
Tray packed without closures; order closures separately (Series 362150).
Note:
Remove cap or completely disengage threads before autoclaving. Autoclave empty. Repeated autoclaving will shorten bottle life.
Bottles shown with colored closures that are available separately.
Specifications
Specifications
| Capacity | 8 oz., 250 mL |
| Capacity (Metric) | 250 mL |
| Capacity (English) | 8 oz. |
| Color | Clear |
| Description | Graduated Square Bottle |
| Diameter (English) | 2.2 in. |
| Diameter (English) Inner Neck | 1.1 in. |
| Diameter (Metric) Inner Neck | 28 mm |
| Material | Polycarbonate |
| No. per Case | 60 |
| Show More |