Learn More
Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes; PP screw closure, silicone gasket
GREENER_CHOICE
Product Code. 10420533
Change view
Click to view available options
For Use With (Equipment):
Replacement O-ring (for Mfr. No. DS3132-0058)
Replacement O-ring (for Mfr. No. DS3132-0063)
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10420533 Replacement O-ring (for Mfr. No. DS3132-0063) Pack of 10
11969436 Replacement O-ring (for Mfr. No. DS3132-0058) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10420533 Supplier Thermo Scientific™ Supplier No. 7131320063
  • ONLINE EXCLUSIVE PRICE
    557.00 NOK
    546.32 NOK / Pack of 10
    Save 10.68 NOK
    1% Off
View all promotions
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
Ensure leakproof service at their maximum speeds >10,000 x g, even when full
  • Polypropylene closures with silicone gasket
Before autoclaving, set closure on top of the container without engaging the threads.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Silicone
Tip Material Silicone
For Use With (Equipment) Replacement O-ring (for Mfr. No. DS3132-0063)