Learn More
Thermo Scientific™ Nalgene™ Repairable PVC Hand-Operated Vacuum Pump Repair Kit
GREENER_CHOICE
Product Code. p-6139465
Change view
Click to view available options
Capacity (Metric):
15 cc
36 cc
No. per Case:
4/Cs.
Unit Size:
Pack of 4
Product Code. Capacity (Metric) No. per Case unitSize
11344535 15 cc 4/Cs. Pack of 4
11354535 36 cc 4/Cs. Pack of 4
2 options
Product Code. 11344535
Brand: Thermo Scientific™ 61322000
Only null left
Add to basket
Add to basket
Everything you need to maintain and repair the internal parts of your Cat. Nos. 6131-series and 6132-series hand pumps for extended hand pump life.
Maintain and repair the internal parts of your Thermo Scientific™ Nalgene™ Cat. Nos. 6131-series and 6132-series hand pumps. Thermo Scientific™ Nalgene™ Repairable Hand-Operated Vacuum Pump Repair Kits include everything you need for extended hand pump life.
Includes: Instructions for maintenance and repair
TRUSTED_SUSTAINABILITY
Tekniske data
Description Repair Kit for Style 15cc PVC Hand Pumps
No. per Case 4/Cs.
Type Vacuum Pump Repair Kit
Capacity (Metric) 15 cc
For Use With Nalgene Vacuum Pumps
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.