Learn More
Thermo Scientific™ Nalgene™ Repairable PVC Hand-Operated Vacuum Pump Repair Kit
GREENER_CHOICE
Product Code. 11344535
Change view
Click to view available options
Capacity (Metric):
15 cc
36 cc
No. per Case:
4/Cs.
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) No. per Case unitSize
11344535 15 cc 4/Cs. Pack of 4
11354535 36 cc 4/Cs. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11344535 Supplier Thermo Scientific™ Supplier No. 61322000
  • ONLINE EXCLUSIVE PRICE
    3835.00 NOK
    3544.73 NOK / Pack of 4
    Save 290.27 NOK
    7% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Everything you need to maintain and repair the internal parts of your Cat. Nos. 6131-series and 6132-series hand pumps for extended hand pump life.
Maintain and repair the internal parts of your Thermo Scientific™ Nalgene™ Cat. Nos. 6131-series and 6132-series hand pumps. Thermo Scientific™ Nalgene™ Repairable Hand-Operated Vacuum Pump Repair Kits include everything you need for extended hand pump life.
Includes: Instructions for maintenance and repair
TRUSTED_SUSTAINABILITY
Specifications
Description Repair Kit for Style 15cc PVC Hand Pumps
No. per Case 4/Cs.
Type Vacuum Pump Repair Kit
Capacity (Metric) 15 cc
For Use With Nalgene Vacuum Pumps