Learn More
Thermo Scientific™ Nalgene™ Polypropylene Graduated Cylinders
GREENER_CHOICE
Product Code. 31352067
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
4000 mL
Graduations:
0.2 mL
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
50 mL
Unit Size:
Each
Product Code. 31352067
Brand: Thermo Scientific™ 36620500
Conduct applications accurately with these graduated cylinders, which feature generous pour spouts and molded-in graduations.
Conduct applications accurately with Thermo Scientific™ Nalgene™ Polypropylene Graduated Cylinders, which feature generous pour spouts, molded-in graduations and a large blue base. There is no meniscus to confuse readings, eliminating guesswork; the cylinders can be chemically sterilized and are suitable for food and beverage use. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:
  • Excellent chemical resistance for a wide variety of solutions
  • Resists temperatures up to 135°C (275°F) for a variety of uses
  • Autoclavable (autoclaving will affect accuracy)
  • Molded-in graduations are silk-screened for easy reading (10mL to 1L sizes only)
Ordering Information:
Individually packaged
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Material Polypropylene
Tolerance ±4 mL
Item Description Graduated Cylinder PP 500mL
Certifications/Compliance ASTM E1272, CFR 21, ISO 6706, Part 177.1520
No. per Pack 1
Calibration To Contain, To Deliver
Diameter (English) 2.44 in.
Scale 15mL to 500mL in 5mL increments
Diameter (Metric) 62 mm
Graduations 5 mL
Autoclavable Autoclavable
Limit of Error ±4.0mL
Max. Temperature 121°C
Shape Round
Diameter (Metric) Inside 46.5 mm
Diameter (English) Inside 1.83 in.
Max. Temperature (English) 250°F
Base Type Hexagonal
Height (English) 15.5 in.
Height (Metric) 393.7 mm
Type Graduated Cylinder
ACT Label Yes