Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks with Closure
Description
Prepare and store culture media using shatter-proof Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks with Closure-- suitable for many culturing techniques. Safer than glass flasks in shakers. Linerless closure protects contents from spills and contamination. Rugged construction combined with light weight make these flasks a valuble element of lab safety programs.
- Molded-in graduations make liquid levels easy to estimate
- Noncontaminating, tight-sealing, linerless polypropylene screw closures protect contents
- Autoclavable/Transparent/Graduated
- Environmental benefits include:
- Renewable Energy
Specifications
Specifications
Capacity (English) | 4 oz. |
Description | Flask Erlenmeyer Screw Closure PC 125mL |
Capacity (Metric) | 125 mL |
Material | Polycarbonate |
No. per Case | 24 |
No. per Pack | 6 |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | Yes |
Mouth Style | Narrow |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Documents
Product Information
Schematics & Diagrams
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
1397820 | Certificate of Compliance | 22/02/2024 | 4108-0125 |
1373126 | Certificate of Compliance | 11/11/2022 | 4108-0125 |
1312210 | Certificate of Compliance | 10/02/2022 | 4108-0125 |
1266441 | Certificate of Compliance | 16/10/2019 | 4108-0125 |
1232287 | Certificate of Compliance | 06/07/2018 | 4108-0125 |