Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
GREENER_CHOICE
Product Code. p-4521533
Change view
Click to view available options
Thread Size (Metric):
20 mm
24 mm
28 mm
38 mm
43 mm
53 mm
63 mm
Product Type:
Replacement Cap
Screw Cap
Spout Cap
Stem and Draw Tube
Color:
Amber
Natural
White
Unit Size:
Pack of 12
Pack of 2
Pack of 25
Product Code. Thread Size (Metric) Product Type Color unitSize
10586551 20 mm Replacement Cap White Pack of 12
10264401 24 mm Replacement Cap Natural Pack of 12
10020661 28 mm Replacement Cap Natural Pack of 12
11334595 28 mm Stem and Draw Tube Natural Pack of 2
10010681 38 mm Replacement Cap Natural Pack of 12
10603452 38 mm Replacement Cap White Pack of 12
10799362 38 mm Screw Cap Natural Pack of 12
10274401 43 mm Replacement Cap Natural Pack of 12
10162491 53 mm Replacement Cap Amber Pack of 12
10000721 53 mm Replacement Cap Natural Pack of 2
10050681 53 mm Replacement Cap Natural Pack of 12
10365941 63 mm Replacement Cap Natural Pack of 12
12716677 20 mm Spout Cap Natural Pack of 25
10644212 24 mm Spout Cap Natural Pack of 25
14 options
Product Code. 10586551
Brand: Thermo Scientific™ 7121510200
Only null left
Add to basket
Add to basket
Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.
Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.
  • Made to fit Nalgene narrow-mouth and wide-mouth bottles
TRUSTED_SUSTAINABILITY
Specifications
Color White
Description Bottle Replacement Closures
Material HDPE
No. per Case 12
Thread Size (Metric) 20 mm
Mouth Narrow
Shape Round
Product Type Replacement Cap
For Use With (Equipment) Nalgene narrow-mouth and wide-mouth bottles
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.