Learn More
Thermo Scientific™ Nalgene™ LDPE Sample Bags
BIOPHARMAGREENER_CHOICE
Product Code. 10126100
Change view
Click to view available options
Length (Metric):
15.2 cm
20.3 cm
33 cm
45.7 cm
Width (Metric):
10.2 cm
12.7 cm
15.2 cm
22.9 cm
Unit Size:
Pack of 250
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) unitSize
10126100 33 cm 15.2 cm Pack of 250
10410981 15.2 cm 10.2 cm Pack of 50
10713461 20.3 cm 12.7 cm Pack of 50
10044620 33 cm 22.9 cm Pack of 50
10626931 45.7 cm 22.9 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Not recommended for storing liquids.
Product Code. 10126100 Supplier Thermo Scientific™ Supplier No. 62550613
Only null left
Add to basket
Add to basket
Not recommended for storing liquids.
Keep dry samples dry and moist samples moist with these LDPE sample bags, ideal for small lab items, botanical samples and other specimens.
Keep dry samples dry and moist samples moist with Thermo Scientific™ Nalgene™ LDPE Sample Bags. Ideal for small lab items, botanical samples and other specimens, these heavy-duty, double-track, low-density polyethylene zipper bags lock securely, but are not recommended for storing liquids.
  • Low-density polyethylene is clear for easy identification of contents
  • 4mil thick walls are strong to protect your samples
  • Bags are waterproof to keep contents dry
  • Transparent
TRUSTED_SUSTAINABILITY
Specifications
Description Heavy-duty double-track zipper bag ideal for small lab items, botanical samples and other specimens
Color Transparent
Material LDPE
Length (English) 13 in.
Width (English) 6 in.
Length (Metric) 33 cm
Width (Metric) 15.2 cm