Learn More
Thermo Scientific™ Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks with Spigots
Product Code. p-4525593
Change view
Click to view available options
Capacity (Metric):
19 L
26.5 L
37.8 L
56.8 L
113.5 L
208 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10305661 19 L Each
10384541 26.5 L Each
10654141 37.8 L Each
10603832 56.8 L Each
10674311 113.5 L Each
10274111 208 L Each
6 options
Product Code. 10305661
Brand: Thermo Scientific™ 111020005
Only null left
Add to basket
Add to basket
Nalgene Heavy-Duty Cylindrical LLDPE Tanks include a needle-type spigot installed for draw off. Spigot accepts 5/8 inch I.D. tubing. Tanks are offered in sizes from 19 to 208L.
Thermo Scientific™ Nalgene Heavy-Duty Cylindrical LLDPE Tanks include a needle-type spigot installed for draw off. This spigot accepts 5/8 inch I.D. tubing and are available in sizes ranging from 19L to 208L.
  • Needle Type Tank Spigot Nalgene (Cat. No. 6421) is installed in tank to allow draw off
  • 5/8 in. (15.9mm) I.D. tubing
  • Offers good temperature and chemical resistance
  • Not autoclavable
Includes:
Tank cover included
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 11 in.
Diameter (Metric) Outer 28 cm
Type Tank with Spigot
Material LLDPE
Capacity (English) 5 gal.
Capacity (Metric) 19 L
Height (English) 15 in.
Height (Metric) 38 cm
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.