Learn More
Thermo Scientific™ Nalgene™ General Long-Term Storage Cryogenic Tubes
BIOPHARMAGREENER_CHOICE
Product Code. p-4526102
Change view
Click to view available options
Capacity (Metric):
1 mL
1.2 mL
1.5 mL
2 mL
2.0 mL
5.0 mL
15 mL
Sterility:
Non-sterile
Sterile
Packaging:
1000 Tubes
250 Tubes
500 Tubes
75 Tubes
Unit Size:
Pack of 1000
Pack of 25
Pack of 250
Pack of 500
Pack of 75
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Sterility Packaging unitSize
11375644 1 mL Sterile 500 Tubes Pack of 500
11740573 1.2 mL Sterile 500 Tubes Pack of 500
11385644 1.5 mL Sterile 500 Tubes Pack of 500
10344691 2.0 mL Sterile 500 Tubes Pack of 25
11750573 2.0 mL Sterile 500 Tubes Pack of 500
11365644 5.0 mL Sterile 250 Tubes Pack of 250
10664321 15 mL Sterile 75 Tubes Pack of 75
10199850 1.2 mL Non-sterile 1000 Tubes Pack of 1000
10543611 2 mL Non-sterile 1000 Tubes Pack of 1000
10662102 1.2 mL Sterile 1000 Tubes Pack of 1000
10731432 2.0 mL Sterile 1000 Tubes Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Do not store in the liquid phase of nitrogen without CryoFlex tubing
Product Code. 11375644 Supplier Thermo Scientific™ Supplier No. 50001012
Only null left
Add to basket
Add to basket
Do not store in the liquid phase of nitrogen without CryoFlex tubing
Enhance aseptic techniques with these externally threaded general long-term storage cryogenic tubes, ideal for research and general lab storage.
Enhance aseptic techniques with externally threaded Thermo Scientific™ Nalgene™ General Long-Term Storage Cryogenic Tubes. These storage tubes are ideally suited for research and general laboratory storage of high-value and genomic materials down to vapor phase liquid nitrogen temperatures.
Features:
  • Large writing areas accommodate more written sample information and graduations for easy visual volume assessment
  • Noncytotoxic raw materials for added sample security and tubes certified free of Dnase and RNase for genomic applications
  • External threading enhances aseptic technique
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • System 100 tubes offer a more dense storage option (10x10) to save precious storage space
  • Self-standing tubes for ease of use on the benchtop
  • Blank, nonsterile and uncapped tubes available in bulk for less demanding applications
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 11.684 mm
Product Type Cryogenic Screw Top
Closure Color Clear
Diameter (English) Outer 0.46 in.
Autoclavable Not Autoclavable
Capacity (Metric) 1 mL
Bottom Shape Conical
Material Polypropylene
Thread Style External
Length (English) 1.5 in.
No. per Case 500
Packaging 500 Tubes
Style SYSTEM 100
RNase Free Yes
Product Line Nalgene
Sterility Sterile
Graduated Yes
Anti-Rotation Yes
Packaging Quantity 500/Cs.
Closure Material Polypropylene
Capacity (English) 0.0338 oz.
Dnase Free Yes
Closure Type Screw Cap
Printed Yes