Learn More
Thermo Scientific™ CryoTube™ Holders
Description
Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.
- Polystyrene coders snap into the caps of Nalgene Cryogenic Tubes for easy sample identification
- Polycarbonate Benchtop Cryogenic Tube Holder has anti-rotation feature for one handed capping and decapping
Specifications
Specifications
| Anti-Rotation | Yes |
| Color | Blue |
| Material | Polycarbonate |
| Type | Vial Holder |
| For Use With (Equipment) | Cryo Tube |
| Holds | 25 Cryogenic Tubes |
| Product Line | Nalgene |
| Quantity | 1/Pk., 4/Cs. |
| Tube Array | 5 x 5 |