Learn More
Thermo Scientific™ CryoTube™ Holders
Product Code. 10100051
Change view
Click to view available options
Holds:
25 Cryogenic Tubes
40 Cryogenic Tubes
50 Cryogenic Tubes
Unit Size:
Each
Pack of 15
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10100051 50 Cryogenic Tubes Each
10508171 25 Cryogenic Tubes Each
10059040 40 Cryogenic Tubes Each
11381665 40 Cryogenic Tubes Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10100051 Supplier Thermo Scientific™ Supplier No. 50300510
  • ONLINE EXCLUSIVE PRICE
    542.00 NOK
    517.68 NOK / Pack of 1
    Save 24.32 NOK
    4% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Increase the functionality of your cryogenic tubes with these benchtop tube and vial holders in a variety of compartment configurations with single-handed capping and decapping.
Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.
  • Polystyrene coders snap into the caps of Nalgene Cryogenic Tubes for easy sample identification
  • Polycarbonate Benchtop Cryogenic Tube Holder has anti-rotation feature for one handed capping and decapping
TRUSTED_SUSTAINABILITY
Specifications
Anti-Rotation Yes
Color White
Material Polycarbonate
Type Vial Holder
For Use With (Equipment) Cryo Tube
Holds 50 Cryogenic Tubes
Product Line Nalgene
Tube Array 5 x 10