Learn More
Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Product Code. p-4530521 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
1 and 1.5 mL Vials
2 mL Vials
5 mL Vials
Vial Capacity:
25
81
100
Quantity:
1/Cs.
10/Cs.
20/Cs.
40/Cs.
80/Cs.
Unit Size:
Pack of 10
Pack of 20
Pack of 25
Pack of 40
Pack of 81
Product Code. Holds Vial Capacity Quantity unitSize
13477649 5 mL Vials 81 1/Cs. Pack of 81
12874372 1 and 1.5 mL Vials 100 10/Cs. Pack of 10
12745918 2 mL Vials 25 80/Cs. Pack of 25
13497649 2 mL Vials 81 40/Cs. Pack of 40
13487649 2 mL Vials 81 20/Cs. Pack of 20
5 options
Product Code. 13477649
Brand: Thermo Scientific™ CS509X10
Only null left
Add to basket
Add to basket
Obtain a comprehensive solution when plastic is preferred to fiberboard.
Obtain a comprehensive solution when plastic is preferred to fiberboard. Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems fit in Thermo Scientific stainless-steel, ultra-low temperature freezer racks.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Type Cryogenic Storage Box
Capacity (Metric) 5 mL
For Use With (Equipment) Locator Jr. Plus, Locator 4 Plus, Locator 6 Plus
Height (English) Exterior 4 in.
Vial Capacity 81
Holds 5 mL Vials
Quantity 1/Cs.