Learn More
Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners, 7 gallon
Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.
Brand:  Thermo Scientific™ 343050-0007
Product Code. 10141133
  • ONLINE EXCLUSIVE PRICE
    7150.00 NOK
    6761.89 NOK / Pack of 10
    Save 388.11 NOK
    5% Off
Only null left
In Stock
Description
Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks. Coex Polyethylene film is free of animal-derived components (ADCF).
  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic