Learn More
Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks
Product Code. 10595971
Change view
Click to view available options
Capacity (Metric):
75 L
115 L
210 L
378.5 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10595971 378.5 L Each
10050461 75 L Each
10338990 115 L Each
10469970 210 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10595971 Supplier Thermo Scientific™ Supplier No. 26500100
  • ONLINE EXCLUSIVE PRICE
    29680.00 NOK
    28077.07 NOK / Pack of 1
    Save 1602.93 NOK
    5% Off
Estimated Shipment: 27-10-2026
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks.
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.
Features:
  • Tanks may be fitted with spigots
  • Autoclavable tanks and closures feature polypropylene construction
Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Comply with FDA Reg. 177.1520 and USP Class VI
Diameter (English) Outer 28.5 in.
Diameter (Metric) Outer 72 cm
Type Closed-dome Tank
Material Polypropylene
Capacity (English) 100 gal.
Capacity (Metric) 378.5 L
Height (English) 52 in.
Height (Metric) 132.1 cm
No. per Case 1
No. per Pack 1
Show More Show Less
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.