Learn More
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Pans
Product Code. 10094740
Change view
Click to view available options
Capacity (Metric):
2 L
2.8 L
3.1 L
5 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10094740 3.1 L Each
10331011 2 L Each
10701601 2.8 L Each
10300391 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10094740 Supplier Thermo Scientific™ Supplier No. 69023000
  • ONLINE EXCLUSIVE PRICE
    827.00 NOK
    760.41 NOK / Pack of 1
    Save 66.59 NOK
    8% Off
Estimated Shipment: 21-12-2026
Repeatedly autoclave instruments and labware with these autoclavable polypropylene pans, featuring heavy-duty construction.
Repeatedly autoclave instruments and labware with Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Pans, featuring heavy-duty construction, extra strong rims and rounded corners for easy cleaning.
  • Tapered for nesting to save space
  • Rolled edges provide extra stiffness for carrying heavy contents
  • Heavy wall construction will withstand rugged use and repeated autoclaving
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 3.1 L
Dimensions (L x W x H) 12.75 x 10.25 x 2.75 in.
Description Polypropylene Autoclavable Pan
Height (Metric) 70 mm
Length (Metric) 324 mm
Material Polypropylene
Type Pan
Width (Metric) 260 mm
Length (English) 12.75 in.
No. per Pack 1
Height (English) 2.75 in.
Shape Rectangular
Width (English) 10.25 in.