missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAGK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15051-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.Specifications
NAGK | |
Polyclonal | |
Unconjugated | |
NAGK | |
GlcNAc kinase, GNK, HSA242910, N acetyl D glucosamine kinase, N acetylglucosamine kinase, NAGK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
297393, 55577, 56174 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P81799, Q9QZ08, Q9UJ70 | |
NAGK | |
NAGK Fusion Protein Ag7034 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |