Description
- Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.
- The square area of 100 mm2 is subdivided into 40 rectangles with an area of 2.5 mm2 each.
- This chamber is commonly used for counting cells in the cerebro-spinal fluid, or for counting nematodes.
Specifications
Specifications
| Product Type | Counting Device |
| Model | Nageotte |
| Quantity | 1Each |
| For Use With (Application) | Counting cells in the cerebro-spinal fluid, or nematodes. |
| Includes | 40 x 2.5mmW rectangles |