missing translation for 'onlineSavingsMsg'
Learn More

NAGA Mouse Monoclonal Antibody, Proteintech

Product Code. 30234504 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
30234504 20 μL 20µL
30233289 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 30234504

Brand: Proteintech 676841IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The NAGA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NAGA. This antibody recognizes human antigen. The NAGA antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen NAGA
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2A5F3
Concentration 500 μg/mL
Conjugate Unconjugated
Gene Symbols D22S674; GALB
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4668
Target Species Human
Content And Storage Store at -20°C.
Antibody Molecular Weight 50 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.