missing translation for 'onlineSavingsMsg'
Learn More

NAF1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27764-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16845815

  • 1415.13 NOK / 20µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RNA-binding protein required for the maturation of box H/ACA snoRNPs complex and ribosome biogenesis. During assembly of the H/ACA snoRNPs complex, it associates with the complex and disappears during maturation of the complex and is replaced by NOLA1/GAR1 to yield mature H/ACA snoRNPs complex. Probably competes with NOLA1/GAR1 for binding with DKC1/NOLA4.
Specifications

Specifications

NAF1
Polyclonal
Unconjugated
NAF1
hNAF1, NAF1
Rabbit
Antigen Affinity Chromatography
RUO
92345
-20°C
Liquid
Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96HR8
NAF1
NAF1 Fusion Protein Ag26787
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers