Learn More
Naepaine Hydrochloride, LoGiCal
Product Code. 30461674
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30461674
Brand: LoGiCal LGCFOR3146.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Naepaine Hydrochloride
CAS 614-42-6
Analyte or Component Names Naepaine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H22 N2 O2 . Cl H
Certifications/Compliance ISO 9001
Synonym Ethanol, 2-(pentylamino)-, 1-(4-aminobenzoate), hydrochloride (1:1), Ethanol, 2-(pentylamino)-, 4-aminobenzoate (ester), monohydrochloride (9CI), Ethanol, 2-(pentylamino)-, p-aminobenzoate (ester), monohydrochloride (8CI), Amylsine hydrochloride, Naepaine hydrochloride
InChI Formula InChI=1S/C14H22N2O2.ClH/c1-2-3-4-9-16-10-11-18-14(17)12-5-7-13(15)8-6-12;/h5-8,16H,2-4,9-11,15H2,1H3;1H
Solution Type Neat
SMILES Cl.CCCCCNCCOC(=O)c1ccc(N)cc1
IUPAC Name 2-(pentylamino)ethyl 4-aminobenzoate;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 286.8
Quantity 10 mg
Formula Weight 286.1448 g/mol