Learn More
Macherey-Nagel™ N9 Polypropylene Yellow Screw Cap
Product Code. 15164722 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Yellow
Unit Size:
Pack of 100
Product Code. Color unitSize
15164722 Yellow Pack of 100
1 options
Product Code. 15164722
Brand: Macherey-Nagel™ 702109
Only null left
Specifications
Color Yellow
Material Polypropylene, PTFE, Silicone
Septum Silicone white / PTFE blue,
slitted
Thickness (Metric) 1 mm
Closure Size N9