Learn More
Honeywell™ N7700 Half Mask Respirator Twin
Product Code. 15846845
Click to view available options
:
N7700 Half Mask Respirator Twin; Size: Small
Product Code. 15846845
Brand: Honeywell™ N65770031
N7700 high performance twin filter half-masks are lightweight with a low profile ergonomic design for a good fit and wearer comfort. The unique cradle headband and yoke piece give ease of adjustment. The contoured revert seal gives an excellent face seal that will not cause discomfort.
  • PPE Category III
  • EN 140, CE Cert 250 - 7700 Respir - Dec 2007
  • 100% silicone hypoallergenic face piece - extra comfort
  • Single unit four-point harness with neck strap - Easy to use, ergonomic design and improved suspension system fits perfectly on top of the head providing a high level of comfort
  • Reduced dead air space - less re-breathed exhaled air
  • Quick to dismantle and clean, with replaceable parts - easy maintenance
  • Superior exhalation valve allows more breath to be exhaled - workers feel cooler and more comfortable
  • Variable thickness in sealing area conforms to nose bridge - improved comfort and fit
Specifications
Certifications/Compliance (Europe) PPE CAT III, EN 140
Certifications (EN) EN 140
Connection Type Threaded Rd40
Color Black
Disposable Reusable
Filter Type Class 1
Product Type Half Mask Respirator
Size Small
With Exhalation Valve Yes
Facepiece Material 100% Silicone
For Use With (Application) Laboratories, pharmaceutical industry - exposure to aerosols containing bacteria or viruses, handling powders, cleanrooms, oil and gas industry - handling chemicals where there is no risk of splashes (gas detection).
Neck Strap Yes
Strap Attachment Type Single unit four-point harness with neck strap