Learn More
N6-methyladenosine (m6A) Antibody (7N6C7) [HRP], Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-33194H
Product Code. 30499838
  • 5880.00 NOK / 0.10mL
Description
N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Non-species specific samples. It is validated for ELISA,Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
N6-methyladenosine (m6A)
Monoclonal
HRP
(-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine
Chemical compounds corresponding to N6-methyladenosine (m6A).
0.1 mL
Primary
Store at 4°C in the dark.
Purified
ELISA, Dot Blot
7N6C7
PBS
Mouse
Affinity purified
RUO
Non-species specific
Antibody
IgG1 κ