Specifications
| Color | White |
| Material | PTFE, Silicone |
| Septum | Silicone White / PTFE Beige |
| For Use With (Application) | Environmental Analysis |
| Thickness (Metric) | 3.2 mm |
| Closure Size | N24 |